Ethyl-2-indolcarboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

OR10188-25G OR10188-5G
APOSOR10188-25GEA 24.8 EUR
APOSOR10188-25G APOSOR10188-5G
Ethyl-2-indolcarboxylat 98%
Ethyl-2-indolcarboxylat
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189.21 g/mol
Schmelzpunkt: 122…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005609
CAS-Nummer: 3770-50-1
EINECS: 223-206-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR