Methyl Indole-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1H-indole-2-carboxylate, Indole-2-carboxylic Acid Methyl Ester

OR10191-100G
APOSOR10191-100GEA 59.6 EUR
APOSOR10191-100G
Methyl Indole-2-carboxylate 97%
Methyl Indole-2-carboxylate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00460779
CAS-Nummer: 1202-04-6
EINECS: 214-861-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR