[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]amine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Aminomethyl)-1,3-dimethyl-1H-pyrazole, (1,3-Dimethyl-1H-pyrazol-5-yl)methanamine

OR101954-1G
APOSOR101954-1GEA 0 EUR
APOSOR101954-1G
[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]amine
[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]amine
Formel: C₆H₁₁N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02682038
CAS-Nummer: 499770-63-7
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR