1-[4-(4-Nitrophenyl)thiazol-2-yl]thiourea
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Amino{[4-(4-nitrophenyl)thiazol-2yl)]amino}methane-1-thione
OR10219-1G
APOSOR10219-1GEA
163
EUR
APOSOR10219-1G
1-[4-(4-Nitrophenyl)thiazol-2-yl]thiourea
1-[4-(4-Nitrophenyl)thiazol-2-yl]thiourea
Formel:
C₁₀H₈N₄O₂S₂ Molecular Weight: 280,326 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00245765 CAS-Nummer: 14015-63-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...