4-Methoxy-2-methylsulphanyl-7H-pyrrolo-(2,3-d)-pyrimidine 95+%

Lieferant: Apollo Scientific

OR1024-1G
APOSOR1024-1GEA 318 EUR
APOSOR1024-1G
4-Methoxy-2-methylsulphanyl-7H-pyrrolo-(2,3-d)-pyrimidine 95+%
4-Methoxy-2-methylsulphanyl-7H-pyrrolo-(2,3-d)-pyrimidine
Formel: C₈H₉N₃OS
Molecular Weight: 195,24 g/mol
Schmelzpunkt: 214…216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02668264
CAS-Nummer: 29877-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR