2-Amino-5-bromo-4,6-dimethylpyrimidine

Lieferant: Apollo Scientific
Achtung

OR10279-500MG
APOSOR10279-500MGEA 21.9 EUR
APOSOR10279-500MG
2-Amino-5-bromo-4,6-dimethylpyrimidine
2-Amino-5-bromo-4,6-dimethylpyrimidine
Formel: C₆H₈BrN₃
Molecular Weight: 202,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00160498
CAS-Nummer: 4214-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR