Dibenzo[b,d]furan-2-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR102860-250MG
APOSOR102860-250MGEA 21.9 EUR
APOSOR102860-250MG
Dibenzo[b,d]furan-2-sulphonyl chloride
Dibenzo[b,d]furan-2-sulphonyl chloride
Formel: C₁₂H₇ClO₃S
Molecular Weight: 266,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00094222
CAS-Nummer: 23602-98-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR