4-(4-Bromophenyl)thiophene-2-carboxylic acid 95+%
Lieferant: Apollo Scientific
OR102870-1G
APOSOR102870-1GEA
241
EUR
APOSOR102870-1G
4-(4-Bromophenyl)thiophene-2-carboxylic acid 95+%
4-(4-Bromophenyl)thiophene-2-carboxylic acid
Formel:
C₁₁H₇BrO₂S Molecular Weight: 283,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407986 CAS-Nummer: 26145-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...