2-(2-Quinoxalinylsulphanyl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: [(Quinoxalin-2-yl)thio]acetic acid

OR102988-1G
APOSOR102988-1GEA 179 EUR
APOSOR102988-1G
2-(2-Quinoxalinylsulphanyl)acetic acid
2-(2-Quinoxalinylsulphanyl)acetic acid
Formel: C₁₀H₈N₂O₂S
Molecular Weight: 220,25 g/mol
Schmelzpunkt: 157…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138691
CAS-Nummer: 69001-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR