5-Amino-2-benzo[b]thiophencarbonsäure

Lieferant: Apollo Scientific

OR103017-1G
APOSOR103017-1GEA 42.2 EUR
APOSOR103017-1G
5-Amino-2-benzo[b]thiophencarbonsäure
5-Amino-2-benzo[b]thiophencarbonsäure
Formel: C₉H₇NO₂S
CAS-Nummer: 98589-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR