3-Acetyl-1-tosylpyrrole 97%

Lieferant: Apollo Scientific

OR10308-25G
APOSOR10308-25GEA 0 EUR
APOSOR10308-25G
3-Acetyl-1-tosylpyrrole 97%
3-Acetyl-1-tosylpyrrole
Formel: C₁₃H₁₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00143087
CAS-Nummer: 106058-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR