4-(1H-Pyrazol-4-yl)pyrimidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR1031-1G
APOSOR1031-1GEA
174
EUR
APOSOR1031-1G
4-(1H-Pyrazol-4-yl)pyrimidine 95+%
4-(1H-Pyrazol-4-yl)pyrimidine
Formel:
C₇H₆N₄ Schmelzpunkt: 205…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02579319 CAS-Nummer: 28648-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...