2-(4-Methoxybenzoyl)indane-1,3-dione 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Methoxybenzoyl)-1H-indene-1,3(2H)-dione, 4-[(2,3-Dihydro-1,3-dioxo-1H-inden-2-yl)carbonyl]anisole

OR1032-1G
APOSOR1032-1GEA 21.9 EUR
APOSOR1032-1G
2-(4-Methoxybenzoyl)indane-1,3-dione 98%
2-(4-Methoxybenzoyl)indane-1,3-dione
Formel: C₁₇H₁₂O₄
Molecular Weight: 280.275 g/mol
Schmelzpunkt: 123…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00129403
CAS-Nummer: 147847-17-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR