4-Carbamoylphenylboronic acid 96%

Lieferant: Apollo Scientific
Achtung

OR10325-25G OR10325-5G OR10325-1G
APOSOR10325-25GEA 0 EUR
APOSOR10325-25G APOSOR10325-5G APOSOR10325-1G
4-Carbamoylphenylboronic acid 96%
4-Carbamoylphenylboronic acid
Formel: C₇H₈BNO₃
Schmelzpunkt: 229…234 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411940
CAS-Nummer: 123088-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR