5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

OR103268-250MG
APOSOR103268-250MGEA 21.9 EUR
APOSOR103268-250MG
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid 96%
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₀H₉N₃O₂
Molecular Weight: 203,2 g/mol
Schmelzpunkt: 183…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00220607
CAS-Nummer: 51649-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR