2-Aminophenylboronsäure Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR10336-1G
APOSOR10336-1GEA 21.9 EUR
APOSOR10336-1G
2-Aminophenylboronsäure Hydrochlorid
2-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173,4052 g/mol
Schmelzpunkt: 145…156 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02258096
CAS-Nummer: 863753-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR