3-Methoxythiophene-2-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methoxythiophene-2-carbaldehyde

OR103362-250MG
APOSOR103362-250MGEA 33.5 EUR
APOSOR103362-250MG
3-Methoxythiophene-2-carboxaldehyde
3-Methoxythiophene-2-carboxaldehyde
Formel: C₆H₆O₂S
Molecular Weight: 142,172 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01571098
CAS-Nummer: 35134-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR