3-Aminophenylboronsäure Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
OR10337-1G
OR10337-5G
OR10337-25G
APOSOR10337-1GEA
0
EUR
APOSOR10337-1G
APOSOR10337-5G
APOSOR10337-25G
3-Aminophenylboronsäure Hydrochlorid
3-Aminophenylboronsäure Hydrochlorid
Formel:
C₆H₉BClNO₂ Schmelzpunkt: >300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00191748 CAS-Nummer: 85006-23-1 EINECS: 285-052-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...