2-Aminophenylboronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
OR10339-25G
APOSOR10339-25GEA
105
EUR
APOSOR10339-25G
2-Aminophenylboronsäurepinakolester
2-Aminophenylboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Schmelzpunkt: 68…70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179448 CAS-Nummer: 191171-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...