4-(1H-Pyrazol-4-yl)quinoline 95+%

Lieferant: Apollo Scientific
Achtung

OR1034-250MG
APOSOR1034-250MGEA 79.9 EUR
APOSOR1034-250MG
4-(1H-Pyrazol-4-yl)quinoline 95+%
4-(1H-Pyrazol-4-yl)quinoline
Formel: C₁₂H₉N₃
Molecular Weight: 195.23 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02579321
CAS-Nummer: 439106-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR