(4-Boc-aminophenyl)boronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR10347-1G OR10347-25G OR10347-5G
APOSOR10347-1GEA 21.9 EUR
APOSOR10347-1G APOSOR10347-25G APOSOR10347-5G
(4-Boc-aminophenyl)boronic acid 98%
(4-Boc-aminophenyl)boronic acid
Formel: C₁₁H₁₆BNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02093054
CAS-Nummer: 380430-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR