2-(1H-Pyrazol-4-yl)quinoxaline 95+%

Lieferant: Apollo Scientific

OR1035-1G OR1035-5G OR1035-10G
APOSOR1035-1GEA 148 EUR
APOSOR1035-1G APOSOR1035-5G APOSOR1035-10G
2-(1H-Pyrazol-4-yl)quinoxaline 95+%
2-(1H-Pyrazol-4-yl)quinoxaline
Formel: C₁₁H₈N₄
Schmelzpunkt: 204…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02579323
CAS-Nummer: 439106-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR