4-Benzyloxy-2-methylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Benzyloxy-2-methylbenzeneboronic acid

OR10356-25G
APOSOR10356-25GEA 206 EUR
APOSOR10356-25G
4-Benzyloxy-2-methylphenylboronsäure
4-Benzyloxy-2-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242.08 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788404
CAS-Nummer: 847560-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR