3-(Benzyloxycarbonyl)phenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR10358-1G OR10358-5G
APOSOR10358-1GEA 24.4 EUR
APOSOR10358-1G APOSOR10358-5G
3-(Benzyloxycarbonyl)phenylboronic acid 98%
3-(Benzyloxycarbonyl)phenylboronic acid
Formel: C₁₄H₁₃BO₄
Schmelzpunkt: 138…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179445
CAS-Nummer: 380430-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR