3-(Benzyloxy)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid
OR10361-1G
APOSOR10361-1GEA
21.9
EUR
APOSOR10361-1G
3-(Benzyloxy)phenylboronsäure
3-(Benzyloxy)phenylboronsäure
Formel:
C₁₃H₁₃BO₃ Molecular Weight: 228,06 g/mol Schmelzpunkt: 125…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093063 CAS-Nummer: 156682-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...