4-(Benzyloxy)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Benzyloxybenzeneboronic acid, (4-(Benzyloxy)phenyl)boronic acid

OR10362-100G OR10362-25G OR10362-5G
APOSOR10362-100GEA 139 EUR
APOSOR10362-100G APOSOR10362-25G APOSOR10362-5G
4-(Benzyloxy)phenylboronsäure 98%
4-(Benzyloxy)phenylboronsäure
Formel: C₁₃H₁₃BO₃
Molecular Weight: 228.06 g/mol
Schmelzpunkt: 198…200 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01075705
CAS-Nummer: 146631-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR