7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-6-carboxypyrazolo[1,5-a]pyrimidine

OR103647-1G OR103647-250MG
APOSOR103647-1GEA 319 EUR
APOSOR103647-1G APOSOR103647-250MG
7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid
7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formel: C₇H₆N₄O₂
Molecular Weight: 178.148 g/mol
Schmelzpunkt: 242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00973861
CAS-Nummer: 68262-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR