1,3-Benzothiazol-2-ylacetic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR103650-20MG
APOSOR103650-20MGEA
21.9
EUR
APOSOR103650-20MG
1,3-Benzothiazol-2-ylacetic acid
1,3-Benzothiazol-2-ylacetic acid
Formel:
C₉H₇NO₂S Schmelzpunkt: 112…113 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00483536 CAS-Nummer: 29182-45-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...