2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Aminomethyl)-1,4-dithiapentalene hydrochloride, (Thieno[3,2-b]thien-2-yl)methylamine hydrochloride
OR103658-1G
OR103658-250MG
OR103658-100MG
APOSOR103658-1GEA
848
EUR
APOSOR103658-1G
APOSOR103658-250MG
APOSOR103658-100MG
2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
Formel:
C₇H₇NS₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702427 CAS-Nummer: 1172699-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...