2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Aminomethyl)-1,4-dithiapentalene hydrochloride, (Thieno[3,2-b]thien-2-yl)methylamine hydrochloride

OR103658-1G OR103658-250MG OR103658-100MG
APOSOR103658-1GEA 848 EUR
APOSOR103658-1G APOSOR103658-250MG APOSOR103658-100MG
2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
2-(Aminomethyl)thieno[3,2-b]thiophene hydrochloride
Formel: C₇H₇NS₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09702427
CAS-Nummer: 1172699-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR