4-Boc(2-aminomethyl)morpholine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate, 2-(Aminomethyl)morpholine, 4-BOC protected

OR103659-1G
APOSOR103659-1GEA 151 EUR
APOSOR103659-1G
4-Boc(2-aminomethyl)morpholine
4-Boc(2-aminomethyl)morpholine
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD06410633
CAS-Nummer: 140645-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR