5-Methylisoxazole-3-carboxamide 97%

Lieferant: Apollo Scientific

OR103676-1G
APOSOR103676-1GEA 43.5 EUR
APOSOR103676-1G
5-Methylisoxazole-3-carboxamide 97%
5-Methylisoxazole-3-carboxamide
Formel: C5H6N2O2
Molecular Weight: 126,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085128
CAS-Nummer: 3445-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR