3-Amino-4-[(4-chlorophenyl)sulphonyl]thiophene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(4-Chlorophenyl)sulphonyl]thiophen-3-amine, 4-Aminothien-3-yl 4-chlorophenyl sulphone

OR103681-250MG
APOSOR103681-250MGEA 140 EUR
APOSOR103681-250MG
3-Amino-4-[(4-chlorophenyl)sulphonyl]thiophene
3-Amino-4-[(4-chlorophenyl)sulphonyl]thiophene
Formel: C₁₀H₈ClNO₂S₂
Molecular Weight: 273,759 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00067899
CAS-Nummer: 175201-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR