3-(Brommethyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Dihydroxy(3-(brommethyl)phenyl)boran, (3-(Brommethyl)phenyl)metaborsäure, 3-(Brommethyl)benzolboronsäure
OR10378-25G
APOSOR10378-25GEA
151
EUR
APOSOR10378-25G
3-(Brommethyl)phenylboronsäure
3-(Brommethyl)phenylboronsäure
Formel:
C₇H₈BBrO₂ Molecular Weight: 214.85 Schmelzpunkt: 216…228 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01632207 CAS-Nummer: 51323-43-4 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...