2-(Bromomethyl)benzeneboronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

OR10379-25G OR10379-5G
APOSOR10379-25GEA 288 EUR
APOSOR10379-25G APOSOR10379-5G
2-(Bromomethyl)benzeneboronic acid pinacol ester
2-(Bromomethyl)benzeneboronic acid pinacol ester
Formel: C₁₃H₁₈BBrO₂
Molecular Weight: 297.00 g/mol
Schmelzpunkt: 78…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632210
CAS-Nummer: 377780-72-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR