4-Bromo-1H-pyrrole-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR10388-100MG
APOSOR10388-100MGEA 21.9 EUR
APOSOR10388-100MG
4-Bromo-1H-pyrrole-2-carbaldehyde
4-Bromo-1H-pyrrole-2-carbaldehyde
Formel: C₅H₄BrNO
Molecular Weight: 174,0000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179586
CAS-Nummer: 931-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR