1-Boc-2-indolboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
OR10393-500MG
APOSOR10393-500MGEA
21.9
EUR
APOSOR10393-500MG
1-Boc-2-indolboronsäure 98%
1-Boc-2-indolboronsäure
Formel:
C₁₃H₁₆BNO₄ Schmelzpunkt: 85…90 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD02093045 CAS-Nummer: 213318-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...