4-(Butylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Butylaminocarbonyl)phenylboronic acid

OR10397-1G
APOSOR10397-1GEA 30.6 EUR
APOSOR10397-1G
4-(Butylcarbamoyl)phenylboronsäure
4-(Butylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03788412
CAS-Nummer: 252663-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR