4-(tert-Butylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR10398-250MG
APOSOR10398-250MGEA 21.9 EUR
APOSOR10398-250MG
4-(tert-Butylcarbamoyl)phenylboronsäure 97%
4-(tert-Butylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Schmelzpunkt: 230…246 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788413
CAS-Nummer: 850568-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR