3-Carboxy-5-nitrophenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid
OR10400-1G
APOSOR10400-1GEA
24.8
EUR
APOSOR10400-1G
3-Carboxy-5-nitrophenylboronsäure
3-Carboxy-5-nitrophenylboronsäure
Formel:
C₇H₆BNO₆ Schmelzpunkt: 229…232 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00757433 CAS-Nummer: 101084-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...