4-Carboxy-2-nitrophenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR10401-250MG
APOSOR10401-250MGEA
21.9
EUR
APOSOR10401-250MG
4-Carboxy-2-nitrophenylboronsäure 97%
4-Carboxy-2-nitrophenylboronsäure
Formel:
C₇H₆BNO₆ Schmelzpunkt: 265…268 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00448780 CAS-Nummer: 85107-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...