2-Chlor-1-methylimidazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-1-methyl-1H-imidazol

OR10413-1G
APOSOR10413-1GEA 104 EUR
APOSOR10413-1G
2-Chlor-1-methylimidazol 95%
2-Chloro-1-methylimidazole
Formel: C₄H₅ClN₂
Molecular Weight: 116,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179531
CAS-Nummer: 253453-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR