2,5-Dichlorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR10444-1G
APOSOR10444-1GEA 21.9 EUR
APOSOR10444-1G
2,5-Dichlorphenylboronsäure
2,5-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
Schmelzpunkt: 210…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863182
CAS-Nummer: 135145-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR