4-(Diethylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid
OR10447-1G
APOSOR10447-1GEA
21.9
EUR
APOSOR10447-1G
4-(Diethylcarbamoyl)phenylboronsäure 98%
4-(Diethylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221,06 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411949 CAS-Nummer: 389621-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...