2,3-Dihydrobenzofuran-5-boronic acid

Lieferant: Apollo Scientific
Achtung

OR10449-5G
APOSOR10449-5GEA 120 EUR
APOSOR10449-5G
2,3-Dihydrobenzofuran-5-boronic acid
2,3-Dihydrobenzofuran-5-boronic acid
Formel: C₈H₉BO₃
Molecular Weight: 163,97 g/mol
Schmelzpunkt: 260…270 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681979
CAS-Nummer: 227305-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR