4,5-Diiodimidazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,5-Diiod-1H-imidazol
OR10450-1G
OR10450-5G
APOSOR10450-1GEA
21.9
EUR
APOSOR10450-1G
APOSOR10450-5G
4,5-Diiodimidazol 98%
4,5-Diiodoimidazole
Formel:
C₃H₂I₂N₂ Schmelzpunkt: 180…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00963878 CAS-Nummer: 15813-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...