4,5-Diiod-2-methylimidazol

Lieferant: Apollo Scientific

OR10453-10G OR10453-1G OR10453-5G
APOSOR10453-10GEA 691 EUR
APOSOR10453-10G APOSOR10453-1G APOSOR10453-5G
4,5-Diiod-2-methylimidazol
4,5-Diiodo-2-methylimidazole
Formel: C₄H₄I₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00464337
CAS-Nummer: 73746-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR