4-(Dimethylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR10459-1G OR10459-25G OR10459-5G
APOSOR10459-1GEA 21.9 EUR
APOSOR10459-1G APOSOR10459-25G APOSOR10459-5G
4-(Dimethylcarbamoyl)phenylboronsäure 97%
Formel: C₉H₁₂BNO₃
Schmelzpunkt: 130…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02258943
CAS-Nummer: 405520-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR