2,3-Dimethylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR10460-500MG
APOSOR10460-500MGEA 21.9 EUR
APOSOR10460-500MG
2,3-Dimethylphenylboronsäure
2,3-Dimethylphenylboronsäure
Formel: C₈H₁₁BO₂
Molecular Weight: 149,98 g/mol
Schmelzpunkt: 174…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863524
CAS-Nummer: 183158-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR