3-(Ethoxycarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

OR10469-25G OR10469-5G OR10469-100G
APOSOR10469-25GEA 34.8 EUR
APOSOR10469-25G APOSOR10469-5G APOSOR10469-100G
3-(Ethoxycarbonyl)phenylboronsäure
3-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
Schmelzpunkt: 131…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179444
CAS-Nummer: 4334-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR